MilliporeSigma
  • Relevance of cognition to health-related quality of life in good-outcome survivors of out-of-hospital cardiac arrest.
Relevance of cognition to health-related quality of life in good-outcome survivors of out-of-hospital cardiac arrest.
Journal of rehabilitation medicine (2015-07-17)
Marte Ørbo, Per M Aslaksen, Kristina Larsby, Christoph Schäfer, Pål M Tande, Torgil R Vangberg, Audny Anke
ABSTRACT
For survivors of out-of-hospital cardiac arrest (OHCA) with good outcomes, it is not known whether and how health-related quality of life is affected by the cognitive impairments frequently observed in these patients. This study explores how neuropsychological tests of memory, exe-cutive and psychomotor functioning relate to the physical and mental aspects of health-related quality of life in functionally independent and community dwelling OHCA survivors discharged early from hospital. The study included 42 adult survivors (mean age 62 years, 38 males). Health-related quality of life was measured approximately 3 months post-OHCA with the Medical Outcome Study Short Form 36 (SF-36). Cognition was measured with established neuropsychological tests. Regression analyses were used to examine associations between neuropsychological domains and physical and mental health-related quality of life, respectively, when controlling for age, education and length of coma. The physical, but not the mental, component of the SF-36 was significantly worse than Norwegian population data. Neuropsychological tests showed frequent impairments most often in the memory domain. Worse psychomotor functioning was associated with worse physical health-related quality of life, whereas worse memory performance was associated with worse mental health-related quality of life. The cognitive impairments frequently reported in OHCA survivors with good outcomes may compromise health-related quality of life. Cognitive functioning should be addressed even in survivors with rapid recovery.
Product Description
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate