MilliporeSigma
  • The equivalence of diffusive samplers to reference methods for monitoring O3, benzene and NO2 in ambient air.
The equivalence of diffusive samplers to reference methods for monitoring O3, benzene and NO2 in ambient air.
Journal of environmental monitoring : JEM (2008-08-30)
Daniela Buzica, Michel Gerboles, Hervé Plaisance
ABSTRACT
A study of the equivalence to the reference methods of the Radiello samplers for ozone (O(3)) and benzene as well as the membrane-closed Palmes tube (MCPT) for nitrogen dioxide (NO(2)) is presented. These samplers benefit from new model equations capable of estimating their uptake rate. For O(3), the aim here was to demonstrate the equivalence for the reference period of 8 h and 120 microg m(-3), the target value of the 3rd European Daughter Directive. For benzene, the demonstration of equivalence to the annual limit value of 5 microg m(-3) of the 2nd European Daughter Directive was examined. In the case of NO(2), the equivalence to the annual limit value of the 1st European Daughter Directive (40 microg m(-3)) was considered. Results show that the radial sampler for O(3) fails to meet the Data Quality Objective (DQO) for continuous monitoring. However, with an expanded uncertainty of less than 30%, the O(3) diffusive sampler fulfils the DQO for indicative measurements. For benzene, the Radiello sampler exposed for 7 days gave satisfying results showing the ability of the sampler to meet the DQO of the reference method. Nevertheless, the field tests should be complemented by measurements for a wider range of benzene concentrations. In the case of NO(2), all the results of the laboratory and field experiments respected the requirements necessary for the demonstration of equivalence. Overall, these findings thus show that the Radiello sampler and the MCPT are equivalent to the reference methods only for assessment of benzene and NO(2), respectively.
Product Description
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello Filtration Kit, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello Hydrogen Sulfide Starter Kit
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Triangular Support Plate, pk of 20
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Barcode Labels, pk of 198
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Outdoor Shelter, pk of 10
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20