MilliporeSigma
A Comparative Study of the Performance of Passive Samplers.
Journal of the Air & Waste Management Association (1995) (2011-03-01)
Bożena Zabiegała, Costel Sărbu, Magdalena Urbanowicz, Jacek Namieśnik
ABSTRACT
Atmospheric concentrations of benzene, toluene, ethylbenzene, and xylenes (o-xylene and m,p-xylene) were assessed in the Tricity area (Gdańsk-Sopot-Gdynia, Poland) with the use of two types of passive samplers: permeation (homemade passive samplers) and diffusive (Radiello and Orsa 5). Samples were collected during 2008 at selected sites in the Tricity area at monitoring stations belonging to the agency of Regional Air Quality Monitoring Foundation. The field study was conducted to compare the performance of these two different types of passive samplers. A statistical approach was formulated, and the experimental data were evaluated using the paired t test, Wilcoxon signed rank-sum test, and Friedman analysis of variance. All the statistical results confirm the hypothesis that the differences between the performances of the three sampling devices are highly significant. Despite the fact that data obtained with the homemade passive sampler indicated that the results were higher compared with those for the Radiello and Orsa 5 diffusive samplers, the authors note that all differences between the homemade permeation sampler and the Radiello and Orsa 5 diffusive samplers are positive. [Box: see text].
Product Description
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Filtration Kit, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Hydrogen Sulfide Starter Kit
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Barcode Labels, pk of 198
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Outdoor Shelter, pk of 10
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Triangular Support Plate, pk of 20