MilliporeSigma
  • Determination of oxathiapiprolin concentration and dissipation in grapes and soil by ultrahigh-performance liquid chromatography-tandem mass spectrometry.
Determination of oxathiapiprolin concentration and dissipation in grapes and soil by ultrahigh-performance liquid chromatography-tandem mass spectrometry.
Journal of the science of food and agriculture (2016-12-16)
Pingzhong Yu, Chunhong Jia, Ercheng Zhao, Li Chen, Hongju He, Junjie Jing, Min He
ABSTRACT
The residue concentrations and dissipation rate of a new oxathiapiprolin fungicide in grapes and soil were investigated to provide an evaluation for the safe use of oxathiapiprolin in grapes. Pesticide residue was extracted by acetonitrile, then purified by solid-phase extraction with an Envi-Carb cartridge and determined by ultrahigh-performance liquid chromatography-tandem mass spectrometry. The fortified recoveries ranged from 85.2% to 99.0%, with relative standard deviations of 1.7-4.5%. The limit of quantitation for oxathiapiprolin in grapes and soil was 0.002 mg kg The residue in the grapes was less than 0.01 mg kg
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS