MilliporeSigma
HomeSupportCalculators & AppsParticle Size Conversion Table
Particle Size Conversion Table
A material’s particle size is typically designated in either microns or mesh. Here you will find a useful particle size conversion table that shows micron to mesh conversions for a fast comparison of particle size between these two units. The chart also allows you to compare sieve designations and sieve openings in inches, millimeters, or microns.
Sieve DesignationNominal Sieve Opening
StandardMeshinchesmmMicrons
25.4 mm1 in.1.0025.425400
22.6 mm7/8 in.0.87522.622600
19.0 mm3/4 in.0.75019.019000
16.0 mm5/8 in.0.62516.016000
13.5 mm0.530 in.0.53013.513500
12.7 mm1/2 in.0.50012.712700
11.2 mm7/16 in.0.43811.211200
9.51 mm3/8 in.0.3759.519510
8.00 mm5/16 in.0.3128.008000
6.73 mm0.265 in.0.2656.736730
6.35 mm1/4 in.0.2506.356350
5.66 mmNo. 3 1/20.2235.665660
4.76 mmNo. 40.1874.764760
4.00 mmNo. 50.1574.004000
3.36 mmNo. 60.1323.363360
2.83 mmNo. 70.1112.832830
2.38 mmNo. 80.09372.382380
2.00 mmNo. 100.07872.002000
1.68 mmNo. 120.06611.681680
1.41 mmNo. 140.05551.411410
1.19 mmNo. 160.04691.191190
1.00 mmNo. 180.03941.001000
0.841 mmNo. 200.03310.841841
0.707 mmNo. 250.02780.707707
0.595 mmNo. 300.02340.595595
0.500 mmNo. 350.01970.500500
0.420 mmNo. 400.01650.420420
0.354 mmNo. 450.01390.354354
0.297 mmNo. 500.01170.297297
0.250 mmNo. 600.00980.250250
0.210 mmNo. 700.00830.210210
0.177 mmNo. 800.00700.177177
0.149 mmNo. 1000.00590.149149
0.125 mmNo. 1200.00490.125125
0.105 mmNo. 1400.00410.105105
0.088 mmNo. 1700.00350.08888
0.074 mmNo. 2000.00290.07474
0.063 mmNo. 2300.00250.06363
0.053 mmNo. 2700.00210.05353
0.044 mmNo. 3250.00170.04444
0.037 mmNo. 4000.00150.03737
Sieve and Mesh Conventions
Large sieve openings (1 in. to 1/4 in.) have been designated by a sieve "mesh" size that corresponds to the size of the opening in inches. Small sieve "mesh" sizes of 3 1/2 to 400 are designated by the number of openings per linear inch in the sieve.
The following convention is used to characterize particle size by mesh designation:
  • "+" before the sieve mesh indicates the particles are retained by the sieve;
  • "-" before the sieve mesh indicates the particles pass through the sieve;
  • typically 90% or more of the particles will lie within the indicated range.
For example, if the particle size of a material is described as -4 +40 mesh, then 90% or more of the material will pass through a 4-mesh sieve (particles smaller than 4.76 mm) and be retained by a 40-mesh sieve (particles larger than 0.420 mm). If a material is described as -40 mesh, then 90% or more of the material will pass through a 40-mesh sieve (particles smaller than 0.420 mm).
Mesh Sieve Products
We offer a variety of mesh screens for cell dissociation and tissue grinding, testing, and particle separation.