Skip to Content
MilliporeSigma
Z-Hyp-OH ≥99.0% (T)

Z-Hyp-OH

Z-Hyp-OH ≥99.0% (T)
Synonym(s):
N-Cbz-Hydroxy-L-proline, Z-L-4-Hydroxyproline
Empirical Formula (Hill Notation):
C13H15NO5
CAS Number:
Molecular Weight:
265.26
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing