Skip to Content
MilliporeSigma
Z-α-Phosphonoglycine trimethyl ester 97%

Z-Phosphonoglycine trimethyl ester

Z-α-Phosphonoglycine trimethyl ester 97%
Synonym(s):
(±)-Z-Phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl) acetate, (±)-Trimethyl Z-phosphonoglycinate
Linear Formula:
(CH3O)2P(O)CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
331.26

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing