Skip to Content
MilliporeSigma
Yttrium(III) trifluoromethanesulfonate 98%

Yttrium(III) trifluoromethanesulfonate

Yttrium(III) trifluoromethanesulfonate 98%
Synonym(s):
Trifluoromethanesulfonic acid yttrium(III) salt, Yttrium triflate
Linear Formula:
(CF3SO3)3Y
CAS Number:
Molecular Weight:
536.11

Products

Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing