Skip to Content
MilliporeSigma
Xylidyl blue I synthetic colorimetric reagent

Xylidyl blue I

Xylidyl blue I synthetic colorimetric reagent
Synonym(s):
4-Hydroxy-3-[2-hydroxy-3-(2,4-dimethylphenylaminocarbonyl)-1-naphthylazobenzenesulfonic acid sodium salt, Magonsulfonate
Linear Formula:
HOC6H3[N=NC10H5(OH)[CONHC6H3(CH3)2]]SO3Na
CAS Number:
Molecular Weight:
513.50
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing