Skip to Content
MilliporeSigma
XPhos Pd G3 98%, 1:1 MTBE adduct

XPhos Pd G3

XPhos Pd G3 98%, 1:1 MTBE adduct
Synonym(s):
(2-Dicyclohexylphosphino-2′,4′,6′-triisopropyl-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate, XPhos-G3-Palladacycle, XPhos-Pd-G3
Empirical Formula (Hill Notation):
C46H62NO3PPdS
CAS Number:
Molecular Weight:
846.45

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing