Skip to Content
MilliporeSigma
Xanthine sodium salt ≥99%

Xanthine sodium salt

Xanthine sodium salt ≥99%
Synonym(s):
2,6-Dihydroxypurine
Empirical Formula (Hill Notation):
C5H3N4NaO2
CAS Number:
Molecular Weight:
174.09

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing