Violanthin
Synonym(s):
6-C-Glucosyl-8-C-rhamnosylapigenin
Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing