Skip to Content
MilliporeSigma
Verteporfin ≥94% (HPLC)

Verteporfin

Verteporfin ≥94% (HPLC)
Synonym(s):
(4R,4aS)-rel-18-ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-Benzo[b]porphine-9,13-dipropanoic acid monomethyl ester, Visudyne, trans-18-ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-23H,25H-Benzo[b]porphine-9,13-dipropanoic acid monomethyl ester
Empirical Formula (Hill Notation):
C41H42N4O8
CAS Number:
Molecular Weight:
718.79

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing