Skip to Content
MilliporeSigma
Venlafaxine Related Compound A certified reference material, pharmaceutical secondary standard

Venlafaxine Related Compound A

Venlafaxine Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
1-[1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol hydrochloride
Empirical Formula (Hill Notation):
C16H25NO2 · HCl
CAS Number:
Molecular Weight:
299.84

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade