Skip to Content
MilliporeSigma
Vanadium(IV) oxide sulfate hydrate ≥99.99% trace metals basis

Vanadium(IV) oxide sulfate hydrate

Vanadium(IV) oxide sulfate hydrate ≥99.99% trace metals basis
Synonym(s):
Vanadyl sulfate hydrate
Linear Formula:
VOSO4 · xH2O
CAS Number:
Molecular Weight:
163.00 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2