Skip to Content
MilliporeSigma
Vanadium(III) acetylacetonate 97%

Vanadium(III) acetylacetonate

Vanadium(III) acetylacetonate 97%
Synonym(s):
2,4-Pentanedione vanadium(III) derivative
Linear Formula:
V(C5H7O2)3
CAS Number:
Molecular Weight:
348.27
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing