![Valganciclovir hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/381/675/8f429106-7ecf-4dbd-a9a2-7882506fc87c/640/8f429106-7ecf-4dbd-a9a2-7882506fc87c.png)
Valganciclovir hydrochloride
![Valganciclovir hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/381/675/8f429106-7ecf-4dbd-a9a2-7882506fc87c/640/8f429106-7ecf-4dbd-a9a2-7882506fc87c.png)
Synonym(s):
L-Valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl ester monohydrochloride
Empirical Formula (Hill Notation):
C14H22N6O5 · HCl
CAS Number:
Molecular Weight:
390.82
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing