Skip to Content
MilliporeSigma
Valeric acid natural, ≥98%, FG

Valeric acid

Valeric acid natural, ≥98%, FG
Synonym(s):
n-Valeric acid, Pentanoic acid
Linear Formula:
CH3(CH2)3COOH
CAS Number:
Molecular Weight:
102.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing