Skip to Content
MilliporeSigma
Uridine 5′-diphosphoglucose disodium salt hydrate from Saccharomyces cerevisiae ≥98%

Uridine 5′-diphosphoglucose disodium salt hydrate from Saccharomyces cerevisiae

Uridine 5′-diphosphoglucose disodium salt hydrate from Saccharomyces cerevisiae ≥98%
Synonym(s):
UDP-Glc hydrate, UDPG hydrate
Empirical Formula (Hill Notation):
C15H22N2Na2O17P2 · xH2O
CAS Number:
Molecular Weight:
610.27 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1