Skip to Content
MilliporeSigma
Ubidecarenone for system suitability European Pharmacopoeia (EP) Reference Standard

Ubidecarenone for System Suitability

Ubidecarenone for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Coenzyme Q10, Q-10, Ubiquinone 50, Ubiquinone-10
Empirical Formula (Hill Notation):
C59H90O4
CAS Number:
Molecular Weight:
863.34

Products

Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique