Skip to Content
MilliporeSigma
UA62784 ≥98% (HPLC)

UA62784

UA62784 ≥98% (HPLC)
Synonym(s):
4-[5-(4-Methoxyphenyl)-2-oxazolyl]-9H-Fluoren-9-one
Empirical Formula (Hill Notation):
C23H15NO3
CAS Number:
Molecular Weight:
353.37

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing