![TWEEN® 20 (Polysorbate) EMPROVE® ESSENTIAL, Ph. Eur., JPE, NF](/deepweb/assets/sigmaaldrich/product/structures/117/277/80f52300-400c-4fe1-88f2-f2dccae30028/640/80f52300-400c-4fe1-88f2-f2dccae30028.png)
TWEEN® 20 (Polysorbate)
![TWEEN® 20 (Polysorbate) EMPROVE® ESSENTIAL, Ph. Eur., JPE, NF](/deepweb/assets/sigmaaldrich/product/structures/117/277/80f52300-400c-4fe1-88f2-f2dccae30028/640/80f52300-400c-4fe1-88f2-f2dccae30028.png)
Synonym(s):
TWEEN® 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate
CAS Number:
Products
Product Category
Brand
Physical Form
Product Line
Agency
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing