Skip to Content
MilliporeSigma
Turkey red oil sodium salt for microscopy

Turkey red oil sodium salt

Turkey red oil sodium salt for microscopy
Synonym(s):
Castor-oil sulfated sodium salt, Sulfated castor oil, Sulforicinolate sodium salt
CAS Number:
EC Number:

Products

Product Category
Brand
pH Value
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing