
TSKgel® Super-Octyl (2.3 µm) HPLC Columns

Synonym(s):
TSKgel® Reversed Phase HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing