Skip to Content
MilliporeSigma
vibrant-m

TSKgel® Phenyl-5PW (13 µm) HPLC Columns

vibrant-m
Synonym(s):
TSKgel® Hydrophobic Interaction Chromatography (HIC) Column

Products

Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1