Skip to Content
MilliporeSigma
TSKgel® GMPWXL HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 13 μm particle size

TSKgel® GMPWXL (13 µm) HPLC Columns

TSKgel® GMPWXL HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 13 μm particle size
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Column

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1