TSKgel® Ether-5PW (10 µm) HPLC Columns
Synonym(s):
TSKgel® Ether-5PW HPLC Column
Products
Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing