Skip to Content
MilliporeSigma
Tropicamide Related Compound A certified reference material, pharmaceutical secondary standard

Tropicamide Related Compound A

Tropicamide Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
4-(Ethylaminomethyl)pyridine, N-(Pyridin-4-ylmethyl)ethanamine, N-Ethyl-4-pyridinemethanamine, NSC 85903
Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade