Tropaeolin O sodium salt
Synonym(s):
4-([2,4-Dihydroxyphenyl]azo)benzenesulfonic acid sodium salt, Acid Orange 6, Chrysoin, Resorcinol yellow
Empirical Formula (Hill Notation):
C12H9N2O5SNa
CAS Number:
Molecular Weight:
316.27
Products
Brand
Formula Weight
pH Value
Chemical Composition
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing