Skip to Content
MilliporeSigma
Trizma® hydrochloride anhydrous, free-flowing, Redi-Dri™, ≥99.0%

Trizma® hydrochloride

Trizma® hydrochloride anhydrous, free-flowing, Redi-Dri™, ≥99.0%
Synonym(s):
TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride
Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
EC Number:

Products

Product Category
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique