Skip to Content
MilliporeSigma
Tris(triphenylphosphine)ruthenium(II) dichloride 97%

Tris(triphenylphosphine)ruthenium(II) dichloride

Tris(triphenylphosphine)ruthenium(II) dichloride 97%
Synonym(s):
Dichlorotris(triphenylphosphine)ruthenium(II), Ruthenium(II)-tris(triphenylphosphine) dichloride
Linear Formula:
[(C6H5)3P]3RuCl2
CAS Number:
Molecular Weight:
958.83
EC Number:

Products

Formula Weight
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing