Skip to Content
MilliporeSigma
Tris(trimethylsilyl) phosphite ≥95.0% (GC)

Tris(trimethylsilyl) phosphite

Tris(trimethylsilyl) phosphite ≥95.0% (GC)
Linear Formula:
[(CH3)3SiO]3P
CAS Number:
Molecular Weight:
298.54

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing