Skip to Content
MilliporeSigma
Tris(trimethylsilyl) borate ≥99.5%, battery grade

Tris(trimethylsilyl) borate

Tris(trimethylsilyl) borate ≥99.5%, battery grade
Linear Formula:
[(CH3)3SiO]3B
CAS Number:
Molecular Weight:
278.38

Products

Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing