Skip to Content
MilliporeSigma
Tris(methylthio)methane 98%

Tris(methylthio)methane

Tris(methylthio)methane 98%
Synonym(s):
Trimethyl trithioorthoformate
Linear Formula:
(CH3S)3CH
CAS Number:
Molecular Weight:
154.32

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing