Skip to Content
MilliporeSigma
Tris(2,2,2-trifluoroethyl) borate 97%

Tris(2,2,2-trifluoroethyl) borate

Tris(2,2,2-trifluoroethyl) borate 97%
Synonym(s):
Tris(2,2,2-trifluoroethyl)borate, Sheppard Reagent
Empirical Formula (Hill Notation):
C6H6BF9O3
CAS Number:
Molecular Weight:
307.91

Products

Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing