Skip to Content
MilliporeSigma
Tri(propylene glycol) butyl ether, mixture of isomers 90%

Tri(propylene glycol) butyl ether, mixture of isomers

Tri(propylene glycol) butyl ether, mixture of isomers 90%
Linear Formula:
CH3(CH2)3O(C3H6O)2C3H6OH
CAS Number:
Molecular Weight:
248.36
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Polymer Composition
Polymer Type
Showing 1-2 of 2