Skip to Content
MilliporeSigma
Triprolidine hydrochloride British Pharmacopoeia (BP) Reference Standard

Triprolidine hydrochloride

Triprolidine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
(E)-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride, trans-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride
Empirical Formula (Hill Notation):
C19H22N2 · HCl
CAS Number:
Molecular Weight:
314.85

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2