Skip to Content
MilliporeSigma
Triphenylphosphine oxide analytical standard

Triphenylphosphine oxide

Triphenylphosphine oxide analytical standard
Synonym(s):
Ph3PO, TPPO, Triphenyl phosphorus oxide, Triphenylphosphine monoxide
Linear Formula:
(C6H5)3PO
CAS Number:
Molecular Weight:
278.28
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reaction Type
Reagent Type
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing