Skip to Content
MilliporeSigma
Tri(o-tolyl)phosphine 97%

Tri(o-tolyl)phosphine

Tri(o-tolyl)phosphine 97%
Synonym(s):
P(o-tol)3, Tris(o-tolyl)phosphine, Tri(o-tolyl)-phosphine
Linear Formula:
(CH3C6H4)3P
CAS Number:
Molecular Weight:
304.37

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing