Skip to Content
MilliporeSigma
Trimipramine maleate European Pharmacopoeia (EP) Reference Standard

Trimipramine maleate

Trimipramine maleate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Trimipramine maleate salt
Empirical Formula (Hill Notation):
C20H26N2 · C4H4O4
CAS Number:
Molecular Weight:
410.51

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade