Skip to Content
MilliporeSigma
Trimethyl(tributylstannyl)silane 97%

Trimethyl(tributylstannyl)silane

Trimethyl(tributylstannyl)silane 97%
Synonym(s):
Tributyl(trimethylsilyl)stannane, (Tributylstannyl)trimethylsilane, (Trimethylsilyl)tributylstannane, (Trimethylsilyl)tributyltin
Linear Formula:
[CH3(CH2)3]3SnSi(CH3)3
CAS Number:
Molecular Weight:
363.24

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing