Skip to Content
MilliporeSigma
Trimethylsilyl trifluoromethanesulfonate purum, ≥98.0% (T)

Trimethylsilyl trifluoromethanesulfonate

Trimethylsilyl trifluoromethanesulfonate purum, ≥98.0% (T)
Synonym(s):
TMS triflate, TMSOTf, Trifluoromethanesulfonic acid trimethylsilylester
Linear Formula:
CF3SO3Si(CH3)3
CAS Number:
Molecular Weight:
222.26
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2