Skip to Content
MilliporeSigma
Trimethyl(propargyl)silane contains 500 ppm BHT as stabilizer

Trimethyl(propargyl)silane

Trimethyl(propargyl)silane contains 500 ppm BHT as stabilizer
Synonym(s):
2-Propynyl-trimethylsilane, 3-(Trimethylsilyl)propyne
Linear Formula:
HC≡CCH2Si(CH3)3
CAS Number:
Molecular Weight:
112.24
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing