Skip to Content
MilliporeSigma
Trimethyl orthobutyrate 97%

Trimethyl orthobutyrate

Trimethyl orthobutyrate 97%
Synonym(s):
1,1,1-Trimethoxybutane
Linear Formula:
CH3CH2CH2C(OCH3)3
CAS Number:
Molecular Weight:
148.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing