Skip to Content
MilliporeSigma
Trimethyl orthoacetate 99%

Trimethyl orthoacetate

Trimethyl orthoacetate 99%
Synonym(s):
1,1,1-Trimethoxyethane, Trimethyl orthoacetate
Linear Formula:
CH3C(OCH3)3
CAS Number:
Molecular Weight:
120.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing