Skip to Content
MilliporeSigma
Triisopropyl borate ≥98%

Triisopropyl borate

Triisopropyl borate ≥98%
Synonym(s):
Boric acid triisopropyl ester, Boron isopropoxide, Triisopropoxyborane, Triisopropyl borate
Linear Formula:
[(CH3)2CHO]3B
CAS Number:
Molecular Weight:
188.07

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing