Skip to Content
MilliporeSigma
Triethylsilyl trifluoromethanesulfonate 99%

Triethylsilyl trifluoromethanesulfonate

Triethylsilyl trifluoromethanesulfonate 99%
Synonym(s):
TES triflate, Trifluoromethanesulfonic acid triethylsilylester
Linear Formula:
CF3SO3Si(C2H5)3
CAS Number:
Molecular Weight:
264.34
EC Number:

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing