Skip to Content
MilliporeSigma
Triethylmethylammonium chloride 97%

Triethylmethylammonium chloride

Triethylmethylammonium chloride 97%
Synonym(s):
Methyltriethylammonium chloride
Linear Formula:
(C2H5)3N(Cl)CH3
CAS Number:
Molecular Weight:
151.68

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing