Skip to Content
MilliporeSigma
Triethylenetetramine dihydrochloride

Triethylenetetramine dihydrochloride

Triethylenetetramine dihydrochloride
Synonym(s):
N,N′-Bis(2-aminoethyl)-1,2-ethanediamine dihydrochloride
Linear Formula:
NH2CH2CH2(NHCH2CH2)2NH2 · 2HCl
CAS Number:
Molecular Weight:
219.16
EC Number:

Products

Product Category
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing