Skip to Content
MilliporeSigma
Trichloro(1H,1H,2H,2H-perfluorooctyl)silane 97%

Trichloro(1H,1H,2H,2H-perfluorooctyl)silane

Trichloro(1H,1H,2H,2H-perfluorooctyl)silane 97%
Synonym(s):
Trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane, 1H,1H,2H,2H-Perfluorooctyl-trichlorosilane
Linear Formula:
CF3(CF2)5CH2CH2SiCl3
CAS Number:
Molecular Weight:
481.54
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing